Skip to Content
Merck
Corning® 384 well plate, clear, poly-D-Lysine, with lid clear polystyrene, flat bottom, non-sterile, lid, case of 100

Corning® 384 well plate, clear, poly-D-Lysine, with lid

Corning® 384 well plate, clear, poly-D-Lysine, with lid clear polystyrene, flat bottom, non-sterile, lid, case of 100
Synonym(s):
Corning® 384 well microplate

Products

Product Category
Color
Manufacturer
Volume (µL)
Wells
Color
Plate Material
Package Size
Sterility