Skip to Content
Merck
Climbazole PESTANAL®, analytical standard

Climbazole

Climbazole PESTANAL®, analytical standard
Synonym(s):
1-(4-Chlorophenoxy)-1-(imidazol-1-yl)-3,3-dimethylbutan-2-one
Empirical Formula (Hill Notation):
C15H17ClN2O2
CAS Number:
Molecular Weight:
292.76
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2