Cibacron Blue 3G-A
Empirical Formula (Hill Notation):
C29H20ClN7O11S3
CAS Number:
Molecular Weight:
774.16
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Chemical Composition
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing