Skip to Content
Merck
vibrant-m

Cetirizine Related Compound A

vibrant-m
Synonym(s):
Cetirizine ethyl ester dihydrochloride, (RS)-2-[2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]acetic acid ethyl ester dihydrochloride
Empirical Formula (Hill Notation):
C23H29ClN2O3 · 2HCl
CAS Number:
Molecular Weight:
489.86

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1