Skip to Content
Merck
Cetirizine hydrochloride Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material

Cetirizine hydrochloride Related Compound A

Cetirizine hydrochloride Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Cetirizine ethyl ester dihydrochloride, (RS)-2-[2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]acetic acid ethyl ester dihydrochloride
Empirical Formula (Hill Notation):
C23H29ClN2O3 · 2HCl
CAS Number:
Molecular Weight:
489.86
EC Number:

Products

Brand
Formula Weight
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique