![Cephapirin Sodium certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/170/694/ae69582a-4cca-4f3c-a680-7bf402064817/640/ae69582a-4cca-4f3c-a680-7bf402064817.png)
Cephapirin sodium
![Cephapirin Sodium certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/170/694/ae69582a-4cca-4f3c-a680-7bf402064817/640/ae69582a-4cca-4f3c-a680-7bf402064817.png)
Synonym(s):
Cefapirin sodium, 3-[(Acetyloxy)methyl]-8-oxo-7-{(4-pyridinylthio)acetyl]amino}-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid monosodium salt, Cephapirin sodium
Empirical Formula (Hill Notation):
C17H16N3NaO6S2
CAS Number:
Molecular Weight:
445.45
Products
Brand
Feature
Formula Weight
Manufacturer
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2