
Cephalothin sodium

Synonym(s):
Cephalothin sodium salt, 7-(2-Thienylacetamido)cephalosporanic acid sodium salt, Cephalotin sodium salt
Empirical Formula (Hill Notation):
C16H15N2NaO6S2
CAS Number:
Molecular Weight:
418.42
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing