Ceftazidime hydrate
Synonym(s):
1-[[(6R,7R)-7-[[(2Z)-(2-amino-4-thiazolyl)[(1-carboxy-1-methylethoxy)imino]acetyl]amino]-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]pyridinum
Empirical Formula (Hill Notation):
C22H22N6O7S2 · xH2O
CAS Number:
Molecular Weight:
546.58 (anhydrous basis)
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Activity Spectrum
Mode of Action
Showing 1-2 of 2
Compare
Product Number
Description
Pricing