![Capsazepine ≥98% (HPLC), solid](/deepweb/assets/sigmaaldrich/product/structures/206/081/7332ceab-4299-49ca-8b82-25b2adef0f38/640/7332ceab-4299-49ca-8b82-25b2adef0f38.png)
Capsazepine
![Capsazepine ≥98% (HPLC), solid](/deepweb/assets/sigmaaldrich/product/structures/206/081/7332ceab-4299-49ca-8b82-25b2adef0f38/640/7332ceab-4299-49ca-8b82-25b2adef0f38.png)
Synonym(s):
N-[2-(4-Chlorophenyl)ethyl]-1,3,4,5-tetrahydro-7,8-dihydroxy-2H-2-benzazepine-2-carbothioamide, Capsazepine
Empirical Formula (Hill Notation):
C19H21ClN2O2S
CAS Number:
Molecular Weight:
376.90
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2