![Canadine phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/289/611/96f04bc0-fc50-425a-aa00-79738f84a5cc/640/96f04bc0-fc50-425a-aa00-79738f84a5cc.png)
Canadine
![Canadine phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/289/611/96f04bc0-fc50-425a-aa00-79738f84a5cc/640/96f04bc0-fc50-425a-aa00-79738f84a5cc.png)
Synonym(s):
Tetrahydroberberine, 5,8,13,13a-Tetrahydro-9,10-dimethoxy-6H-benzo[g]-1,3-benzodioxolo[5,6-alpha]quinolizine, Canadine, THB, Tetrahydroumbellatine, Xanthopuccine
Empirical Formula (Hill Notation):
C20H21NO4
CAS Number:
Molecular Weight:
339.39
Products
Formula Weight
Manufacturer
Product Line
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing