Skip to Content
Merck
Calpain Inhibitor I ≥97% (TLC), powder

Calpain Inhibitor I

Calpain Inhibitor I ≥97% (TLC), powder
Synonym(s):
Calpain Inhibitor I, ALLN, Ac-LLnL-CHO, MG-101, N-Acetyl-L-leucyl-L-leucyl-L-norleucinal, N-Acetyl-Leu-Leu-Norleu-al
Empirical Formula (Hill Notation):
C20H37N3O4
CAS Number:
Molecular Weight:
383.53

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2