![Calcium phosphate dried, EMPROVE® ESSENTIAL, Ph. Eur., BP, E 341 (iii)](/deepweb/assets/sigmaaldrich/product/images/311/048/1468862b-2c09-408f-810c-26967205badf/640/1468862b-2c09-408f-810c-26967205badf.jpg)
Calcium phosphate
![Calcium phosphate dried, EMPROVE® ESSENTIAL, Ph. Eur., BP, E 341 (iii)](/deepweb/assets/sigmaaldrich/product/images/311/048/1468862b-2c09-408f-810c-26967205badf/640/1468862b-2c09-408f-810c-26967205badf.jpg)
Synonym(s):
Calcium phosphate
Empirical Formula (Hill Notation):
Ca3O8P2
CAS Number:
Molecular Weight:
310.18
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing