![β-Sitosterol D-glucoside phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/231/321/56df0eca-a24d-4c2c-87d2-71391689cdb6/640/56df0eca-a24d-4c2c-87d2-71391689cdb6.png)
β-Sitosterol D-glucoside
![β-Sitosterol D-glucoside phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/231/321/56df0eca-a24d-4c2c-87d2-71391689cdb6/640/56df0eca-a24d-4c2c-87d2-71391689cdb6.png)
Synonym(s):
β-Sitosterol β-D-glucoside, β-Daucosterol, Coriandrinol, Daucosterin, Eleutheroside A
Empirical Formula (Hill Notation):
C35H60O6
CAS Number:
Molecular Weight:
576.85
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Optical Isomer
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing