
Boc-Arg(Tos)-OH

Synonym(s):
Nα-Boc-Nω-tosyl-L-arginine, Boc-Arg(Tos)-OH
Empirical Formula (Hill Notation):
C18H28N4O6S
CAS Number:
Molecular Weight:
428.50
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing