
Bismuth subsalicylate

Synonym(s):
Bismuth(III) salicylate basic
Linear Formula:
HOC6H4COOBiO
CAS Number:
Molecular Weight:
362.09
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing