Biotinamidohexanoic acid N-hydroxysuccinimide ester
Synonym(s):
(+)-Biotinamidocaproate N-hydroxysuccinimidyl ester, N-(+)-Biotinyl-6-aminocaproic acid N-succinimidyl ester, N-(+)-Biotinyl-6-aminohexanoic acid NHS ester, N-Succinimidyl N-biotinyl-6-aminocaproate, Succinimidyl 6-(biotinamido)hexanoate
Empirical Formula (Hill Notation):
C20H30N4O6S
CAS Number:
Molecular Weight:
454.54
Products
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing