Skip to Content
Merck
β-Endorphin human ≥95% (HPLC)

β-Endorphin human

β-Endorphin human ≥95% (HPLC)
Synonym(s):
β-Lipotropin 61-91, β-Lipotropin C-fragment, LPH (61-91)
Empirical Formula (Hill Notation):
C158H251N39O46S
CAS Number:
Molecular Weight:
3464.98

Products

Product Category
Brand
Formula Weight
Gene Alias
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing