Skip to Content
Merck
vibrant-m

β-Acetyldigoxin

vibrant-m
Synonym(s):
Digoxin 4″′-acetate
Empirical Formula (Hill Notation):
C43H66O15
CAS Number:
Molecular Weight:
822.98

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing