Anti-USP2 antibody produced in rabbit
Synonym(s):
Anti-41 kDa ubiquitin-specific protease antibody produced in rabbit, Anti-Deubiquitinating enzyme 2 antibody produced in rabbit, Anti-Ubiquitin carboxyl-terminal hydrolase 2 antibody produced in rabbit, Anti-Ubiquitin thioesterase 2 antibody produced in rabbit, Anti-Ubiquitin-specific-processing protease 2 antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing