
Anti-TFRC antibody produced in rabbit

Synonym(s):
Anti-CD71, Anti-TFR, Anti-TFR1, Anti-TR, Anti-TRFR, Anti-Transferrin receptor (p90, CD71), Anti-Transferrin receptor protein 1, Anti-p90, AntiT9
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2