![Anti-ALDH18A1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/255/266/22d9ec16-3158-4522-8b6d-05d59a0daec8/640/22d9ec16-3158-4522-8b6d-05d59a0daec8.jpg)
Anti-ALDH18A1 antibody produced in rabbit
![Anti-ALDH18A1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/255/266/22d9ec16-3158-4522-8b6d-05d59a0daec8/640/22d9ec16-3158-4522-8b6d-05d59a0daec8.jpg)
Synonym(s):
Anti-Δ1-pyrroline-5-carboxylate synthetase antibody produced in rabbit, Anti-Aldehyde dehydrogenase 18 family member A1 antibody produced in rabbit, Anti-P5CS antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing