Skip to Content
Merck
Anastrozole Pharmaceutical Secondary Standard; Certified Reference Material

Anastrozole

Anastrozole Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
2;2"-[5-(1H-1;2;4-Triazol-1-ylmethyl)-1, 3-Phenylene]bis(2-methyl-propiononitrile, Arimidex, ICI-D1033, ZD1033, a,a,a′,a′-Tetramethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-1,3-benzenediacetonitrile, Anastrozole
Empirical Formula (Hill Notation):
C17H19N5
CAS Number:
Molecular Weight:
293.37

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique