
Amrinone Related Compound B

Synonym(s):
N-[1,6-Dihydro-6-oxo(3,4′-bipyridine)-5-yl]-2-hydroxypropanamide
Empirical Formula (Hill Notation):
C13H13N3O3
Molecular Weight:
259.26
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2