![Amisulpride British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/733/831/06ba9661-e0a3-4fb3-941a-8cd34b78560f/640/06ba9661-e0a3-4fb3-941a-8cd34b78560f.png)
Amisulpride
![Amisulpride British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/733/831/06ba9661-e0a3-4fb3-941a-8cd34b78560f/640/06ba9661-e0a3-4fb3-941a-8cd34b78560f.png)
Synonym(s):
4-Amino-N-[(1-ethyl-2-pyrrolidinyl)methyl]-5-(ethylsulfonyl)-2-methoxybenzamide, DAN-2163, Deniban
Empirical Formula (Hill Notation):
C17H27N3O4S
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3