all-cis-7,10,13,16,19-Docosapentaenoic acid
Empirical Formula (Hill Notation):
C22H34O2
CAS Number:
Molecular Weight:
330.50
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing