![Albumin human recombinant, expressed in Pichia pastoris, lyophilized powder, ≥90% (agarose gel electrophoresis)](/deepweb/assets/sigmaaldrich/product/images/172/595/2f0e7ca4-0040-4ee6-929f-146d77f02743/640/2f0e7ca4-0040-4ee6-929f-146d77f02743.jpg)
Albumin human
![Albumin human recombinant, expressed in Pichia pastoris, lyophilized powder, ≥90% (agarose gel electrophoresis)](/deepweb/assets/sigmaaldrich/product/images/172/595/2f0e7ca4-0040-4ee6-929f-146d77f02743/640/2f0e7ca4-0040-4ee6-929f-146d77f02743.jpg)
Synonym(s):
rHSA
Empirical Formula (Hill Notation):
C2936H4624N786O889S41
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Recombinant Host
Quality Segment
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing