Skip to Content
Merck
6-Methyl-5-hepten-2-one natural, ≥98%, FCC

6-Methyl-5-hepten-2-one

6-Methyl-5-hepten-2-one natural, ≥98%, FCC
Linear Formula:
(CH3)2C=CHCH2CH2COCH3
CAS Number:
Molecular Weight:
126.20
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing