Skip to Content
Merck
5-Aminopyridine-2-carboxylic acid 96%

5-Aminopyridine-2-carboxylic acid

5-Aminopyridine-2-carboxylic acid 96%
Synonym(s):
5-amino-2-pyridinecarboxylic acid, 5-Amino-2-picolinic acid
Empirical Formula (Hill Notation):
C6H6N2O2
CAS Number:
Molecular Weight:
138.12

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing