Skip to Content
Merck
4-Ethylguaiacol analytical standard

4-Ethylguaiacol

4-Ethylguaiacol analytical standard
Synonym(s):
4-Ethyl-2-methoxyphenol, NSC 82313
Linear Formula:
C2H5C6H3-2-(OCH3)OH
CAS Number:
Molecular Weight:
152.19
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing