4-Benzyloxy-1-butanol
Synonym(s):
Tetramethylene glycol monobenzyl ether
Linear Formula:
C6H5CH2O(CH2)4OH
CAS Number:
Molecular Weight:
180.24
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing