
4-Allyloxy-2-hydroxybenzophenone

Linear Formula:
H2C=CHCH2OC6H3(OH)COC6H5
CAS Number:
Molecular Weight:
254.28
Products
Brand
Formula Weight
Melting Point (°C)
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing