Skip to Content
Merck
3-Phenylpropionic acid 99%, FG

3-Phenylpropionic acid

3-Phenylpropionic acid 99%, FG
Synonym(s):
Hydrocinnamic acid, 3-Phenylpropionic acid, Benzylacetic acid
Linear Formula:
C6H5CH2CH2COOH
CAS Number:
Molecular Weight:
150.17
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing