
2-Oxoadipic acid

Synonym(s):
α-Ketoadipic acid, 2-Oxoadipate, 2-Oxohexanedioic acid
Empirical Formula (Hill Notation):
C6H8O5
CAS Number:
Molecular Weight:
160.12
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1