2-Hydroxyibuprofen
Synonym(s):
2-[4-(2-Hydroxy-2-methylpropyl)phenyl]propionic acid
Empirical Formula (Hill Notation):
C13H18O3
CAS Number:
Molecular Weight:
222.28
Products
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing