Skip to Content
Merck
2-Azabicyclo[2.2.1]hept-5-en-3-one 98%

2-Azabicyclo[2.2.1]hept-5-en-3-one

2-Azabicyclo[2.2.1]hept-5-en-3-one 98%
Synonym(s):
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 4-Amino-2-cyclopentene-1-carboxylic acid lactam
Empirical Formula (Hill Notation):
C6H7NO
CAS Number:
Molecular Weight:
109.13

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing