
2-(4-tert-Butyl-3-hydroxy-2,6-dimethylphenyl)acetonitrile

Synonym(s):
2-[4-(1,1-Dimethylethyl)-3-hydroxy-2,6-dimethylphenyl]acetonitrile
Empirical Formula (Hill Notation):
C14H19NO
CAS Number:
Molecular Weight:
217.31
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1