![2,4-Disulfamyl-5-trifluoromethylaniline United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/361/847/97259211-8051-4187-afcc-b582af6ee227/640/97259211-8051-4187-afcc-b582af6ee227.png)
2,4-Disulfamyl-5-trifluoromethylaniline
![2,4-Disulfamyl-5-trifluoromethylaniline United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/361/847/97259211-8051-4187-afcc-b582af6ee227/640/97259211-8051-4187-afcc-b582af6ee227.png)
Synonym(s):
4-Amino-6-(trifluoromethyl)benzene-1,3-disulfonamide, 5-Amino-α,α,α-trifluorotoluene-2,4-disulfonamide
Empirical Formula (Hill Notation):
C7H8F3N3O4S2
CAS Number:
Molecular Weight:
319.28
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing