Skip to Content
Merck
2-(2-Butoxyethoxy)ethyl acetate ≥99.2%

2-(2-Butoxyethoxy)ethyl acetate

2-(2-Butoxyethoxy)ethyl acetate ≥99.2%
Synonym(s):
Butyldiglycol acetate, Diethylene glycol monobutyl ether acetate, [2-(2-Butoxyethoxy)ethyl] acetate
Linear Formula:
CH3COOCH2CH2OCH2CH2O(CH2)3CH3
CAS Number:
Molecular Weight:
204.26

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing