Skip to Content
Merck
2,2′-Bipyridine-5,5′-dicarboxylic acid 97%

2,2′-Bipyridine-5,5′-dicarboxylic acid

2,2′-Bipyridine-5,5′-dicarboxylic acid 97%
Synonym(s):
6,6′-Binicotinic acid
Empirical Formula (Hill Notation):
C12H8N2O4
CAS Number:
Molecular Weight:
244.20

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing