1-Iodobutane
Synonym(s):
Butyl iodide, 1-Iodobutane
Linear Formula:
CH3(CH2)3I
CAS Number:
Molecular Weight:
184.02
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing