
1H,1H,2H,2H-Perfluoro-1-dodecanol

Synonym(s):
3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-Heneicosafluoro-1-dodecanol
Empirical Formula (Hill Notation):
C12H5F21O
CAS Number:
Molecular Weight:
564.13
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Reference Material Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing