
(12-Phosphonododecyl)phosphonic acid

Empirical Formula (Hill Notation):
C12H28O6P2
CAS Number:
Molecular Weight:
330.29
Products
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing