Purge and Trap
The purge and trap technique purges inert gas through the sample containing volatile organic compounds (VOCs) into an adsorbent trap. The VOCs are then desorbed from the trap by heating and transferring the analyte to the GC column. With minimal sample handling, this technique ensures reliable and reproducible results.
The purge and trap method is ideal for extracting and concentrating VOCs from water, soil, and sludges. The adsorbent filled trap (purge trap) allows the VOCs from the sample to be concentrated while managing the water vapor during the purge step or the desorb step (instrument dependent) to achieve low ppb (parts per billion) detection limits.
This technique is routinely utilized for environmental monitoring, analysis of flavors in food, and toxic compounds in bodily fluids.
Related Product Resources
- Brochure: Gas Chromatography - Tools for Your Workflow
We provide a comprehensive range of high quality tools for each step of the GC workflow ensuring accuracy, precision and consistency of your analysis.
Purge Traps
For purge and trap analysis, the tubes are usually packed with multiple beds of adsorbent materials to trap a wide range of polar, non-polar, high, and low molecular weight compounds in a single tube. Each bed in the trap protects the succeeding active bed by preventing the strong adsorption of compounds.
Glassware and Accessories for Purge and Trap Analysis
We offer a variety of accessories and glassware for purge and trap analysis, including female Luer lock connectors, fritless purge samplers, sample valves, Hamilton® syringes, Thermogreen® LB-1 septa, male Luer lock connectors, gas sampling bulbs, PTFE Stopcock plugs, needle sparge glassware, and needle purge tubes.