Skip to Content
Merck
All Photos(1)

Key Documents

Z744266

Wilmad® Suprasil® Aqueous Cell, Spectroscopic Quality

volume 150 μL, pack of 1 ea

Synonym(s):

EPR aqueous cell, aqueous cell

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15

packaging

pack of 1 ea

manufacturer/tradename

Wilmad WG-812-S-Q

volume

150 μL

General description

When the sample contains water, the electric dipole moment of the water molecule interacts with the E field in the resonator and a large amount of microwave energy is absorbed by the sample. Flat cells confine the sample to a 0.3 mm space to solve this issue. The hexagonal stems are available. The sample volume is 150 μl for all cells. Suprasil® is used in the critical portions of the cell. This cell has optically polished flat windows are joined to the polished wall ends via a special thermal fusion technique to form an ultra high precision bore flat cell. The two windows are parallel to each other within several visible light wave numbers. These items are recommended for EPR experiments that requires critical coupling.

Application

EPR

Features and Benefits

0.3 mm Sample space prevents the absorbtion of microwave energy.

Legal Information

Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Wilmad is a registered trademark of SP Industries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service