Skip to Content
Merck
All Photos(1)

Key Documents

BLO-11

90112702, mouse muscle, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

BLO-11, 90112702

biological source

mouse muscle

growth mode

Adherent

karyotype

2n = 40

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Moblo/y muscle hereditary emphysema

Cell Line Description

Established from the abdominal wall muscle of a 30 day old male Blotchy mouse (Mo (blo/y)) with hereditary emphysema. The cells are deficient in lysyl oxidase activity and accumulate high levels of intracellular copper. NOR-10 (Sigma Catalogue number. 90112701) was isolated from the abdominal wall muscle of a normal litter mate of the mouse from which BLO-11 was derived, thus it can serve as a matched control for studies with BLO fibroblasts.

Application

Genetic studies

Culture Medium

DMEM + 2mM Glutamine + 20% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:3 i.e. seeding at 3-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service