Skip to Content
Merck
All Photos(1)

Documents

52048

Sigma-Aldrich

Hexaethylene glycol monotetradecyl ether

BioXtra, ≥99.0% (GC)

Synonym(s):

C14E6, Tetradecylhexaglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13(OCH2CH2)6OH
CAS Number:
Molecular Weight:
478.70
Beilstein:
1805220
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

product line

BioXtra

Assay

≥99.0% (GC)

form

solid

mol wt

478.70 g/mol

technique(s)

RT-PCR: suitable
protein expression: suitable
protein purification: suitable
western blot: suitable

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cr: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Mn: ≤50 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

CCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C26H54O7/c1-2-3-4-5-6-7-8-9-10-11-12-13-15-28-17-19-30-21-23-32-25-26-33-24-22-31-20-18-29-16-14-27/h27H,2-26H2,1H3

InChI key

CMOAVXMJUDBIST-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G C Terstappen et al.
Biotechnology and applied biochemistry, 16(3), 228-235 (1992-12-01)
The partitioning of a variety of extracellular lipases, both pro- and eucaryotic, in detergent-based aqueous two-phase systems was examined. The results revealed that all procaryotic lipases showed a clear preference for the detergent-rich coacervate phase. In contrast, all eucaryotic lipases
A Schwarz et al.
Analytical biochemistry, 254(2), 221-225 (1998-01-07)
A rapid, simple, and efficient chloroform/methanol-free method of isolating gangliosides is described. The nonionic polyoxyethylene detergent, hexaethyleneglycol mono-n-tetradecyl ether (C14EO6), forms clear micellar solutions in water, but two-phase separation can be achieved by centrifugation in the presence of ammonium sulfate
Cloud-point extraction of gangliosides using nonionic detergent C14EO6.
G C Terstappen et al.
Methods in enzymology, 312, 187-196 (2000-11-09)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service