Skip to Content
Merck
All Photos(2)

Key Documents

86546

Supelco

Dibenzyl chloromethyl phosphate

for HPLC derivatisation, LiChropur, ≥97.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16ClO4P
CAS Number:
Molecular Weight:
326.71
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for HPLC derivatization

Quality Level

Assay

≥97.0% (HPLC)

form

liquid

quality

LiChropur

shelf life

limited shelf life, expiry date on the label

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

technique(s)

HPLC: suitable

storage temp.

−20°C

SMILES string

ClCOP(=O)(OCc1ccccc1)OCc2ccccc2

InChI

1S/C15H16ClO4P/c16-13-20-21(17,18-11-14-7-3-1-4-8-14)19-12-15-9-5-2-6-10-15/h1-10H,11-13H2

InChI key

FVTHQGOGQRTOMY-UHFFFAOYSA-N

Application

Synthesis of water-soluble prodrugs of lipophilic alcohols, phenols and amines. This compound is an improved derivative of the well known Di-tert-butyl chloromethyl phosphate. So it can be detected more easily by HPLC-UV, because of higher stability, higher yields and better UV-activity.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel synthetic route for the preparation of alkyl and benzyl chloromethyl phosphates
Mantyla, A., et al.
Tetrahedron Letters, 43(21), 3793-3794 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service