Skip to Content
Merck
All Photos(1)

Key Documents

74178

Sigma-Aldrich

1-Allylpiperazine

purum, ≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14N2
CAS Number:
Molecular Weight:
126.20
MDL number:
UNSPSC Code:
12352100

grade

purum

Assay

≥97.0% (GC)

impurities

≤0.5% water

refractive index

n20/D 1.4830 (lit.)
n20/D 1.483

bp

181-182 °C (lit.)

density

0.902 g/mL (lit.)

SMILES string

C=CCN1CCNCC1

InChI

1S/C7H14N2/c1-2-5-9-6-3-8-4-7-9/h2,8H,1,3-7H2

InChI key

ZWAQJGHGPPDZSF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service