Skip to Content
Merck
All Photos(1)

Key Documents

MABE433

Sigma-Aldrich

Anti-phospho Histone H2B (Tyr37), clone 15 Antibody

clone 15, from mouse

Synonym(s):

Histone H2B type 3-B, H2B type 12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

15, monoclonal

species reactivity

human, mouse

technique(s)

ChIP: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr37)

Gene Information

human ... H2BC1(255626)
mouse ... H2Bc1(319177)

General description

Histones are basic nuclear proteins that are responsible for the nucleosome structure of chromatin in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which DNA is wrapped in repeating units, called nucleosomes. Histone H2B is one of the main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H2B is involved with the structure of the nucleosomes of the ′beads on a string′ structure.

Immunogen

KLH-conjugated linear peptide corresponding to human Histone H2B phosphorylated at Tyr37.

Application

Chromatin Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated phospho Histone H2B (Tyr37) in mouse MEF cells which were untreated or treated with WEE inhibitor (Mahajan, K., et al. (2012). Nat Struct Mol Biol. 19(9):930-937.).

Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated phospho Histone H2B (Tyr37) in MEF-overexpressing WEE (Mahajan, K., et al. (2012). Nat Struct Mol Biol. 19(9):930-937.).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
This Anti-phospho Histone H2B (Tyr37) antibody, clone 15 is validated for use in western blotting & ChIP for the detection of phospho Histone H2B (Tyr37).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected phospho Histone H2B (Tyr37) in 10 µg of HeLa cell lysate.

Target description

~17 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service