Skip to Content
Merck
All Photos(2)

Key Documents

MABC1143

Sigma-Aldrich

Anti-MED28 Antibody, clone 7E1

clone 7E1, from mouse

Synonym(s):

Mediator of RNA polymerase II transcription subunit 28, Endothelial-derived protein 1, Magicin, Mediator complex subunit 28, Merlin and Grb2-interacting cytoskeletal protein, Tumor angiogenesis marker EG-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€322.00

€322.00


Estimated to ship on23 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€322.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

€322.00


Estimated to ship on23 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7E1, monoclonal

species reactivity

human, rat, monkey, mouse

technique(s)

electron microscopy: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... MED28(80306)

General description

Mediator of RNA polymerase II transcription subunit 28 (UniProt Q9H204; also known as Endothelial-derived protein 1, Magicin, Mediator complex subunit 28, Merlin and Grb2-interacting cytoskeletal protein, Tumor angiogenesis marker EG-1) is encoded by the MED28 (also known as EG1, FKSG20) gene (Gene ID 80306) in human. MED28 (Magicin) is a subunit of the Mediator complex that is composed of at least 30 subunits and plays an essential role in transcriptional regulation by bridging RNA polymerase II transcriptional machinery and DNA-bound regulatory factors. MED28 is reported to mediate cell surface receptor-mediated signaling and associate with Src-family kinases (e.g. Fyn, Src and Lck), as well as with the Neurofibromatosis 2 (NF2) tumor suppressor merlin. MED28 is associated with both the cytoskeleton and the nucleus, indicating a role as an adaptor/scaffolding protein to relay cellular signals to the cytoskeleton and the nucleus. Homozygous Med28-deletion causes peri-implantation lethality with a loss of pluripotency within the inner cell mass (ICM) of the blastocyst. In addition, Med28-/- embryonic fibroblasts (MEFs) lose the capability to reprogram and Med28 overexpression augments reprogramming efficiency. Upregulated MED28 expression is associated with poor outcome in breast cancer patients, and targeted MED28 downregulation decreases breast cancer cell proliferation and tumor growth.

Specificity

Clone 7E1 detected MED28 in wild-type, but not Med28-/- murine cells. Clone 7E1 detected Cre expression-induced MED28 knockdown in MEFs with floxed Med28 alleles (Li, L., et al. (2015). PLoS One. 10(10):e0140192).

Immunogen

GST-tagged recombinant human MED28 lacking N-terminal 12 amino acids.

Application

Detect MED28 using this mouse monoclonal Anti-MED28 Antibody, clone 7E1, Cat. No. MABC1143, validated for use in Electron Microscopy, Immunocytochemistry, and Western Blotting.
Research Category
Apoptosis & Cancer
Western Blotting Analysis: 1 µg/mL from a representative lot detected MED28 in 10 µg of human embryonic kidney HEK293 cell lysates.

Electron Microscopy Analysis: A representative lot detected MED28 (Magicin) immunoreactivity on cortical actin filaments, colocalizing with that of merlin by immunogold staining of 1% glutaraldehyde-fixed, 0.1% SDS-treated CAD mouse neuronal cells (Wiederhold, T., et al. (2004). Oncogene. 23(54):8815-8825).

Immunocytochemistry Analysis: A representative lot immunostained 4% paraformaldehyde-fixed, 0.25% Triton X-100-permeabilized wild-type, but not Med28-/-, mouse blastocysts, as well as wild-type MEF-dervied iPSC colonies (Li, L., et al. (2015). PLoS One. 10(10):e0140192).

Immunocytochemistry Analysis: A representative lot detected MED28 (Magicin) immunoreactivity colocalized with that of phalloidin-stained actin filament by flourescent immunocytochemistry staining of 0.1% SDS-treated CAD mouse neuronal cells (Wiederhold, T., et al. (2004). Oncogene. 23(54):8815-8825).

Western Blotting Analysis: A representative lot detected a higher MED28 expression in mouse embryonic stem cells (ESCs) than in embryonic fibroblasts (MEFs). Clone 7E1 detected Cre expression-induced MED28 knockdown in MEFs with floxed Med28 alleles (Li, L., et al. (2015). PLoS One. 10(10):e0140192).

Western Blotting Analysis: A representative lot detected ~24 kDa MED28 (Magicin) in human (F293, Gli238, HEK293T, HeLa, MeT5A, T-47D), monkey (COS-7), and mouse (CAD) cell lysates, as well as in Merlin immunoprecipitates from T-47D cytosolic/heavy membrane fractions (Cyt/HMF) and whole cell lysate (Wiederhold, T., et al. (2004). Oncogene. 23(54):8815-8825).

Quality

Evaluated by Western Blotting in RAW267.7 murine macrophage lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected MED28 in 10 µg of RAW267.7 murine macrophage lysate.

Target description

~20 kDa observed. 19.52 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service